Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

tri-Sodium citrate 3.8% (w/v) in aqueous solution

Supplier: Ricca Chemical

R7228000-500A R7228000-120A
RC72284EA 37.31 USD InStock
RC72284 RC722816
tri-Sodium citrate 3.8% (w/v) in aqueous solution
tri-Sodium citrate solutions


Formula: Na₃C₆H₅O₇·2H₂O
MW: 294.1 g/mol
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.0250 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00150031
CAS Number: 6132-04-3
Order Now
Specification Test Results
Appearance Clear, colorless, odorless
Assay 3.8 ± 0.3 % w/v

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty