Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium diethyldithiocarbamate 0.1% (w/v) in aqueous solution

Supplier: Ricca Chemical

R7240600-500A
RC7240.6-16EA 41.01 USD InStock
RC7240.6-16
Sodium diethyldithiocarbamate 0.1% (w/v) in aqueous solution
Sodium diethyldithiocarbamate


Formula: C₅H₁₀NNaS₂
MW: 171.26 g/mol
CAS Number: 148-18-5
UN: 3077
ADR: 9,III
Order Now
Specification Test Results
Appearance Clear, colorless, odorless
Suitability for Use Yellow-brown color with Copper

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty