Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium cyanide 2.5% (w/v) in aqueous solution

Supplier: Ricca Chemical

R7241000-120A
RC72414EA 51.8 USD InStock
RC72414
Sodium cyanide 2.5% (w/v) in aqueous solution
Sodium cyanide


Formula: CNNa
MW: 49.01 g/mol
MDL Number: MFCD00003523
CAS Number: 143-33-9
UN: 1689
ADR: 6.1,I
Order Now
Specification Test Results
Appearance Clear, colorless, cyanide odor

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty