Synonyms:
Caustic sodaLyeSodium hydrateSoda lye
R7280000-1A
R7280000-20F
R7280000-500A
R7280000-4A
RCR7280000500AEA
247.4
USD
InStock
RCR7280000500A
RC72805
RCR7280000-1A
RC72801
Sodium hydroxide 40% (w/v) in aqueous solution for Kjeldahl nitrogen analysis nitrogen-free
Sodium hydroxide
Nitrogen-free, suitable for Kjeldahl nitrogen tests.