Synonyms:
Sodium hydrate
, Soda lye
, Caustic soda
, Lye
R7290000-15E
R7290000-20F
R7290000-55E
R7290000-4A
R7290000-20E1
R7290000-1A
RC72905EA
580.11
USD
InStock
RC72905
RC7290-5HP
RCR7290000-15E
RCR7290000-55E
RC72901
RCR7290000-1A
Sodium hydroxide 50% (w/w) in aqueous solution (760 g/L) for Kjeldahl nitrogen analysis nitrogen-free, purified
Sodium hydroxide
Nitrogen-free, suitable for Kjeldahl nitrogen tests.