Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide 50% (w/w) in aqueous solution (760 g/L) analytical reagent

Supplier: Ricca Chemical
Danger
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

R7291000-500A R7291000-30E R7291000-20E1 R7291000-20F R7291000-4A
RC72915EA 1256.12 USD InStock
RC72915 RC7291-5HP RCR7291000-30E RC729116 RC72911
Sodium hydroxide 50% (w/w) in aqueous solution (760 g/L) analytical reagent
Sodium hydroxide


Formula: NaOH
MW: 40 g/mol
Boiling Pt: ~100 °C
Melting Pt: 12 °C
Density: 1.53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II
Order Now
Specification Test Results
Appearance Colorless liquid
Assay at 20 °C (traceable to NIST SRM 723) 50 - 52 % (w/w)
Chloride (Cl) 0.002% max
Heavy Metals (as Ag) 10 ppm (0.001%) max
Iron (Fe) 5 ppm (0.0005%) max
NH4OH Precipitate 0.01% max
Nickel (Ni) 5 ppm (0.0005%) max
Nitrogen Compounds (as N) 5 ppm (0.0005%) max
Phosphate (PO4) 5 ppm (0.0005%) max
Potassium (K) 0.01% max

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty