Synonyms:
Caustic sodaLyeSodium hydrateSoda lye
R7300000-20F
R7300000-500A
R7300000-4A
R7300000-1A
RCR7300000500AEA
33.39
USD
InStock
RCR7300000500A
RCR7300000-20F
RC73001
RC730032
Sodium hydroxide 0.0200 N (N/50) (1 ml = 1 mg CaCO₃)
Sodium hydroxide
APHA for acidity.