Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide 0.100 N (N/10) in aqueous solution

Supplier: Ricca Chemical
Synonyms: Caustic sodaLyeSodium hydrateSoda lye

R7350000-20F R7350000-4A R7350000-10F R7350000-1A R7350000-500A
RC735032CS 585.58 USD InStock
RC735032 RC735025 RC73501 RC735016 RC73505
Sodium hydroxide 0.100 N (N/10) in aqueous solution
Sodium hydroxide
Standardized at 25°C against chemicals whose certification is traceable to NIST.


Formula: NaOH
MW: 40 g/mol
Density: 1 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,III
Order Now
Specification Test Results
Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 84) 0.1000 ± 0.0001 N at 20.0 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty