Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide 0.1142 N

Supplier: Ricca Chemical
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

R7355000-4A R7355000-20F
RC73551EA 95.13 USD InStock
RC73551 RC73555
Sodium hydroxide 0.1142 N
Sodium hydroxide


Formula: NaOH
MW: 40 g/mol
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1823
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 84) 0.1142 ± 0.0001 N at 20.0 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty