Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide 0.250 N (N/4)

Supplier: Ricca Chemical
Warning
Synonyms: Caustic sodaLyeSodium hydrateSoda lye

R7370000-20F R7370000-500A R7370000-10F R7370000-4A R7370000-1A
RC737032EA 54.76 USD InStock
RC737032 RC7370-2.5 RC73701 RCR7370000500A RC73705
Sodium hydroxide 0.250 N (N/4)
Sodium hydroxide
Clear, colorless liquid.


Formula: NaOH
MW: 40 g/mol
Boiling Pt: ∼105 °C (1013 hPa)
Melting Pt: 0.0°C
Density: 1.01
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: UN1824
ADR: 8,III
Order Now
Specification Test Results
Appearance Colorless liquid
Assay (vs. Potassium Hydrogen Phthalate/Phenolphthalein) 0.2499-0.2501 N at 20°C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty