Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide - EDTA solution 10 N

Supplier: Ricca Chemical
Danger

R7478000-1A R7478000-500A
RC7478-32EA 150.17 USD InStock
RC7478-32 RC7478-16
Sodium hydroxide - EDTA solution 10 N
Sodium hydroxide - EDTA solution


Boiling Pt: ~ 100 °C
Density: 1.4000 g/cm³
Storage Temperature: Ambient
UN: 1824
ADR: 8,II
Order Now
Specification Test Results
Ammonia Electrode No detectable Ammonia
Appearance Clear, colorless, alcohol odor
Assay at 20 °C (traceable to NIST SRM 723) 10.0 ± 0.1 N at 20.0 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty