Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ionic strength adjustor (ISA) Sodium Nitrate 5.0 M

Supplier: Ricca Chemical

R7496000-500A
RC749616EA 151.75 USD InStock
RC749616
Ionic strength adjustor (ISA) Sodium Nitrate 5.0 M
Ionic strength adjustor (ISA)


Storage Temperature: Ambient
UN: 1498
ADR: 5.1,III
Order Now
Specification Test Results
Appearance Clear, colorless, odorless
Assay 5.00 ± 0.07 M at 20.0 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty