Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium nitroprusside 0.5 g/L in aqueous solution

Supplier: Ricca Chemical

R7498000-1C R7498000-500C
RC7498-32EA 122.62 USD InStock
RC7498-32 RC7498-16
Sodium nitroprusside 0.5 g/L in aqueous solution
Sodium nitroprusside


Formula: C₅FeN₆Na₂O
MW: 261.92 g/mol
MDL Number: MFCD00003506
CAS Number: 14402-89-2
UN: 1588
ADR: 6.1,II
Order Now
Specification Test Results
Appearance Clear, tan color, odorless

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty