Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium phenoxide solution for ammonia nitrogen determination

Supplier: Ricca Chemical
Danger
Synonyms: Sodium phenateSodium benzenolateSodium phenylate

R7518000-500A R7518000-1A
RC7518-16EA 101.56 USD InStock
RC7518-16 RC7518-32
Sodium phenoxide solution for ammonia nitrogen determination
Sodium phenoxide


Formula: C₆H₅NaO
MW: 116.09 g/mol
Boiling Pt: ~ 100 °C
Density: 1.1000 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00013134
CAS Number: 139-02-6
UN: 3267
ADR: 8,III
Order Now
Specification Test Results
Appearance Clear, tan to reddish, phenol odor
Suitability for Use Passes Test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty