Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium sulfate 22.2% (w/v) in aqueous solution

Supplier: Ricca Chemical
Synonyms: Sulphuric acid disodium saltBisodium sulphateSulfuric acid disodium saltBisodium sulfateDisodium sulphateDisodium sulfate

R7800000-500A R7800000-1A
RC780032EA 82.35 USD InStock
RC780032 RC780016
Sodium sulfate 22.2% (w/v) in aqueous solution
Sodium sulphate


Formula: Na₂SO₄
MW: 142.04 g/mol
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.0050 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003504
CAS Number: 7757-82-6
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/41/16640641.jpg
Order Now
Specification Test Results
Appearance Clear, colorless, odorless
Refractive Index 22.2 ± 0.5 % (w/v)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty