Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium tetraphenylborate USP Test Solution (TS)

Supplier: Ricca Chemical
Synonyms: Sodium tetraphenylboronTetraphenyl sodium borateTetraphenylboron sodium salt

R7854000-500C
RC7854-16EA 143.27 USD InStock
RC7854-16
Sodium tetraphenylborate USP Test Solution (TS)
Sodium tetraphenylborate


Formula: (C₆H₅)₄BNa
MW: 342.22 g/mol
MDL Number: MFCD00011494
CAS Number: 143-66-8
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Clear, colorless, odorless

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty