Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium thiocyanate 0.100 N (N/10) in aqueous solution

Supplier: Ricca Chemical
Warning
Synonyms: Sodium rhodanide

R7860000-1A R7860000-10F R7860000-4A R7860000-20F
RC786025EA 200.03 USD InStock
RC786025 RC7860-1 RC7860-5 RC7860-32
Sodium thiocyanate 0.100 N (N/10) in aqueous solution
Sodium thiocyanate


Formula: NaSCN
MW: 81.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011123
CAS Number: 540-72-7
EINECS: 231-867-5
Order Now
Specification Test Results
Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 999) 0.1000 ± 0.0001 N at 20.0 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty