Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium tungstate 10% (w/v) in aqueous solution stabilized

Supplier: Ricca Chemical
Warning

R7980000-500A
RC798016EA 73.59 USD InStock
RC798016
Sodium tungstate 10% (w/v) in aqueous solution stabilized
Sodium tungstate solutions


Formula: Na₂WO₄·2H₂O
MW: 329.85 g/mol
Boiling Pt: ~ 100 °C
Density: 1.0900 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00149190
CAS Number: 10213-10-2
Order Now
Specification Test Results
Appearance Clear, colorless, odorless
Specific Gravity 1.09 ± 0.02 sp gr

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty