Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Strong acid solution for phosphorus determination

Supplier: Ricca Chemical
Danger

R8080000-500A
RC808016EA 54.84 USD InStock
RC808016
Strong acid solution for phosphorus determination
Strong acid solution


Boiling Pt: ~ 103 °C
Density: 1.3200 g/cm³
Storage Temperature: Ambient
UN: 3264
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear, colorless, odorless

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty