Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sulfanilamide reagent 1% (w/v) in hydrochloric acid 10%

Supplier: Ricca Chemical
Danger
Synonyms: 4-Aminobenzenesulfonamide4-Aminobenzenesulphonamide

R8103000-500A R8103000-1A
RC810316EA 89.9 USD InStock
RC810316 RC8103-32
Sulfanilamide reagent 1% (w/v) in hydrochloric acid 10%
Sulphanilamide


Formula: H₂NC₆H₄SO₂NH₂
MW: 172.21 g/mol
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.0200 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00007939
CAS Number: 63-74-1
UN: 1789
ADR: 8,III
Order Now
Specification Test Results
Appearance Clear, colorless, odorless

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty