Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Sulfosalicylic acid 20% (w/v) in aqueous solution

Supplier: Ricca Chemical
Danger
Synonyms: 2-Hydroxy-5-sulphobenzoic acid2-Hydroxy-5-sulfobenzoic acid

R8130000-4C R8130000-500C R8130000-1C
RC8130-1EA 447.79 USD InStock
RC8130-1 RC813016 RC813032
5-Sulfosalicylic acid 20% (w/v) in aqueous solution
5-Sulfosalicylic acid solutions


Formula: HO₃SC₆H₃-2-(OH)CO₂H·2H₂O
Boiling Pt: ~ 100 °C
Density: 1.1000 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00007508
CAS Number: 5965-83-3
Order Now
Specification Test Results
Appearance Clear, tannish-gold to light pink, odorless
Assay at 20 °C (traceable to NIST SRM 84) 20.00 ± 0.02 % (w/v)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty