Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Silver sulfate solution in sulfuric acid for COD determination

Supplier: Ricca Chemical
Danger
Synonyms: Disilver sulphateSilver(I) sulfateDisilver sulfate

R8350000-1C1 R8350000-500C1
RC835016EA 269.44 USD InStock
RC835016 RC835032
Silver sulfate solution in sulfuric acid for COD determination
Silver sulphate


Formula: Ag₂SO₄
MW: 311.8 g/mol
Boiling Pt: ~ 338 °C
Density: 1.8000 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003407
CAS Number: 10294-26-5
UN: 1830
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear, colorless, odorless

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty