Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tannic acid 25 g/ 100 mL in water for direct count for bacteria, yeast, and mold

Supplier: Ricca Chemical
Synonyms: TanninGallotannin

R8368000-4A R8368000-1A
RC8368-1EA 710.61 USD InStock
RC8368-1 RC8368-32
Tannic acid 25 g/ 100 mL in water for direct count for bacteria, yeast, and mold
Tannic acid
Contains Sodium Benzoate


Formula: C₇₆H₅₂O₄₆
MW: 1701.2 g/mol
Boiling Pt: ~ 100 °C
Density: 1.0897 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00066397
CAS Number: 1401-55-4
Order Now
Specification Test Results
Appearance Clear, brown

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty