Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

TCLP extraction fluid 1 for use in EPA 1311 toxicity characteristic leaching procedure pH 4.93 ± 0.05 (25°C)

Supplier: Ricca Chemical
Danger

R8377000-4A R8377000-20F
RC83771EA 127.45 USD InStock
RC83771 RC83775
TCLP extraction fluid 1 for use in EPA 1311 toxicity characteristic leaching procedure pH 4.93 ± 0.05 (25°C)
TCLP extraction fluid 1
pH 4.93 ± 0.05 at 25°C.


Order Now
Specification Test Results
Appearance Clear, colorless, slight Acetic odor
pH at 25 °C (traceable to NIST SRM 185 & 186) 4.93 ± 0.02 pH at 25.0 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty