Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Thioacetamide 0.25% (w/v) in aqueous solution

Supplier: Ricca Chemical
Danger
Synonyms: Ethanethioamide

R8393000-120A R8393000-500A
RC839316EA 84.26 USD InStock
RC839316 RC83934
Thioacetamide 0.25% (w/v) in aqueous solution
Thioacetamide


Formula: CH₃CSNH₂
MW: 75.13 g/mol
Boiling Pt: 100 °C
Melting Pt: 0.0 °C
Density: 1.0010 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00008070
CAS Number: 62-55-5
https://us-legacy.vwr.com/stibo/web/std.lang.all/72/66/2297266.jpg
Order Now
Specification Test Results
Appearance Clear, colorless, odorless to slight mercaptan odor

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty