Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Thymol blue 0.4% (w/v) in aqueous solution indicator

Supplier: Ricca Chemical
Synonyms: ThymolsulphonphthaleinThymolsulfonphthalein3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

R8410000-500A R8410000-120A
RC841016EA 115.26 USD InStock
RC841016 RC84104
Thymol blue 0.4% (w/v) in aqueous solution indicator
Thymol blue


Formula: C₂₇H₃₀O₅S
MW: 466.6 g/mol
Boiling Pt: 100 °C
Melting Pt: 0.0 °C
Density: 1.0010 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00005869
CAS Number: 76-61-9
Order Now
Specification Test Results
Appearance Dark green, odorless
Suitability for Use Red (pH 9.2)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty