Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Thymol blue 0.1% (w/v) in alcohol USP Test Solution (TS), USP Reference Solution (RS)

Supplier: Ricca Chemical
Danger
Synonyms: ThymolsulphonphthaleinThymolsulfonphthalein3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

R8422000-500A R8422000-120A
RC8422-4EA 105.73 USD InStock
RC8422-4 RC8422-16
Thymol blue 0.1% (w/v) in alcohol USP Test Solution (TS), USP Reference Solution (RS)
Thymol blue


Formula: C₂₇H₃₀O₅S
MW: 466.6 g/mol
Boiling Pt: ~ 77 °C
Density: 0.8000 g/cm³
Flash Pt: ~ 15.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00005869
CAS Number: 76-61-9
UN: 1987
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear, purplish-red, alcohol odor
Suitability for Use Red (pH 9.2)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty