Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Thymolphthalein 0.1% (w/v) in alcohol

Supplier: Ricca Chemical
Danger
Synonyms: 3,3-Bis(4-hydroxy-5-isopropyl-o-tolyl)phthalide

R8460000-120A R8460000-500A
RC8460-4EA 81.64 USD InStock
RC8460-4 RC8460-16
Thymolphthalein 0.1% (w/v) in alcohol
Thymolphthaleine


Formula: C₂₈H₃₀O₄
MW: 430.54 g/mol
Boiling Pt: ~ 77 °C
Density: 0.8000 g/cm³
Flash Pt: ~ 15.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00005909
CAS Number: 125-20-2
UN: 1987
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear, colorless, alcohol odor
Suitability for Use Colorless (pH 10.5)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty