Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Barium chloride 12% (w/v) in aqueous solution USP Test Solution (TS), USP Reference Solution (RS)

Supplier: Ricca Chemical

R0854000-500A R0854000-1A
RC854-32EA 87.18 USD InStock
RC854-32 RC854-16
Barium chloride 12% (w/v) in aqueous solution USP Test Solution (TS), USP Reference Solution (RS)
Barium chloride


Formula: BaCl₂
MW: 208.24 g/mol
MDL Number: MFCD00003445
CAS Number: 10361-37-2
UN: 1564
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Clear, colorless, odorless
Assay 12.0 ± 0.3 % (w/v)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty