Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Picric acid 1% (w/v) in aqueous solution TS

Supplier: Ricca Chemical
Warning

R8696800-500A
RC8696.8-16EA 140.21 USD InStock
RC8696.8-16
Picric acid 1% (w/v) in aqueous solution TS
Picric acid


Formula: C₆H₃N₃O₇
MW: 229.11 g/mol
Boiling Pt: 100 °C
Melting Pt: 0.0 °C
Density: 1.0010 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00007102
CAS Number: 88-89-1
Order Now
Specification Test Results
Appearance Clear, yellow, odorless

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty