Would you like to log in again?
Turk's solution, diluting fluid for leukocyte count
Supplier: Ricca Chemical
Danger
R8850000-500A R8850000-1A
RC885032EA 64.99 USD InStock
RC885032 RC885016
Turk's solution, diluting fluid for leukocyte count
Turk's solution
For In Vitro Diagnostic Use. Suitable for use as a diluting fluid for Leukocyte (white blood cell) counting.
IVDLogo
Order Now
Specification Test Results
Absorbance (traceable to NIST SRM 930) Conforms to control
Appearance Purple, odorless
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty