Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

pH Indicator solution universal pH 2-10

Supplier: Ricca Chemical
Danger

R8870000-500C R8870000-1C R8870000-120C
RC8870-4EA 82.27 USD InStock
RC8870-4 RC8870-32 RC8870-16
pH Indicator solution universal pH 2-10
pH Indicator solution


Boiling Pt: ~ 77 °C
Density: 0.8000 g/cm³
Flash Pt: 13 °C
Storage Temperature: Ambient
UN: 1987
ADR: 3,II
Order Now
Specification Test Results
Appearance Yellow, odorless
Suitability for Use Color Changes OK

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty