Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Barium hydroxide saturated solution in water USP Test Solution (TS) pH 12.9

Supplier: Ricca Chemical

R0898000-1A
RC898-32EA 154.18 USD InStock
RC898-32
Barium hydroxide saturated solution in water USP Test Solution (TS) pH 12.9
Barium hydroxide


Formula: H₂BaO₂
MW: 171.34 g/mol
MDL Number: MFCD00003443
CAS Number: 17194-00-2
UN: 3262
ADR: 8,III
Order Now
Specification Test Results
Appearance Slightly hazy, colorless, odorless

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty