Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bathocuproine 0.1% (w/v) in aqueous solution

Supplier: Ricca Chemical

R0905000-500A R0905000-120A
RC90516EA 916.37 USD InStock
RC90516 RC9054
Bathocuproine 0.1% (w/v) in aqueous solution
Bathocuproine


Boiling Pt: 100 °C
Melting Pt: 0.0 °C
Density: 1.0000 g/cm³
Storage Temperature: Ambient
CAS Number: 52698-84-7
Order Now
Specification Test Results
Appearance Clear, colorless, no odor

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty