Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Eriochrome Black T 0.5% (w/v) in 2-methoxyethanol for hardness of water determinations

Supplier: Ricca Chemical
Synonyms: Sodium 3-hydroxy-4-(2-hydroxy-1-naphthylazo)-7-nitro-1-naphthalenesulphonate Solochrome Black T EBT

Eriochrome®
R9300000-500C R9300000-120C
RC93004EA 38.88 USD InStock
RC93004 RC930016
Eriochrome Black T 0.5% (w/v) in 2-methoxyethanol for hardness of water determinations
Eriochrome Black T
In 2-Methoxyethanol.


Formula: C₂₀H₁₂N₃NaO₇S
MW: 461.39 g/mol
MDL Number: MFCD00003935
CAS Number: 1787-61-7
UN: 3077
ADR: 9,III
Order Now
Specification Test Results
Appearance Dark red, Methoxyethanol odor
Suitability for Use Wine Red (excess Ca or Mg) - Pure Blue (excess EDTA)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty