Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Benedict's reagent quantitative

Supplier: Ricca Chemical

935-16
RC935-16EA 77.12 USD InStock
RC935-16
Benedict's reagent quantitative
Benedict's reagent


MDL Number: MFCD00146068
CAS Number: 63126-89-6
UN: 3082
ADR: 9,III
Order Now
Specification Test Results
Assay 2 ± 0.01 mg Glucose/ mL

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty