Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Wright's stain for staining and general differentiation of blood corpuscles, regular formula

Supplier: Ricca Chemical
Danger
Synonyms: Eosin Methylene blue solution according to WrightWright's eosin methylene blue

R9360000-4C R9360000-500C R9360000-1C
RC93601EA 355.42 USD InStock
RC93601 RC936016 RC936032
Wright's stain for staining and general differentiation of blood corpuscles, regular formula
Wright's stain


Boiling Pt: ~ 64.5 °C
Density: 0.8000 g/cm³
Flash Pt: ~ 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00082143
CAS Number: 68988-92-1
UN: 1993
ADR: 3,II
Order Now
Specification Test Results
Absorbance (traceable to NIST SRM 930) Conforms to control
Appearance Dark blue, Methanol odor
pH at 25 °C (traceable to NIST SRM 185 & 186) 6.4 ± 0.1 pH at 25.0 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty