Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Wright-Giemsa stain

Supplier: Ricca Chemical
Danger
Synonyms: Wright-Giemsa Solution

R9380000-500C R9380000-1C
RC938016EA 95.31 USD InStock
RC938016 RC938032
Wright-Giemsa stain
Wright-Giemsa stain (solutions & kits)
Wright-Giemsa Stain Mixture


Boiling Pt: ~ 64.5 °C
Density: 0.8484 g/cm³
Flash Pt: ~ 12 °C (MeOH)
Storage Temperature: Ambient
MDL Number: MFCD00132593
UN: 1993
ADR: 3,II
Order Now
Specification Test Results
Absorbance (traceable to NIST SRM 930) Conforms to control
Appearance Dark blue, Methanol odor

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty