Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Xylenol orange 0.1% (w/v) in alcohol USP Test Solution (TS)

Supplier: Ricca Chemical
Danger

R9387000-120A R9387000-500A
RC9387-16EA 238.88 USD InStock
RC9387-16 RC9387-4
Xylenol orange 0.1% (w/v) in alcohol USP Test Solution (TS)
Xylenol orange


Formula: C₃₁H₂₈N₂Na₄O₁₃S
MW: 760.59 g/mol
MDL Number: MFCD00044293
CAS Number: 3618-43-7
UN: 1987
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear, red, alcohol odor
Suitability for Use Purple (Excess Metal) - Yellow (Excess EDTA)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty