Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Zinc acetate 2 N

Supplier: Ricca Chemical

R9450000-120A R9450000-500A
RC94504EA 37.86 USD InStock
RC94504 RC945016
Zinc acetate 2 N
Zinc acetate
220 g + 870 ml H₂O


Formula: C₄H₆O₄Zn
MW: 183.48 g/mol
MDL Number: MFCD00012454
CAS Number: 557-34-6
UN: 3077
ADR: 9,III
Order Now
Specification Test Results
Appearance Clear, colorless, odorless
Refractive Index Confirmed concentration

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty