Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Zinc sulfate 1.180 Specific Gravity for concentration of intestinal parasites, eggs, and cysts

Supplier: Ricca Chemical

R9650000-1A R9650000-4A R9650000-500A
RC96501EA 232.38 USD InStock
RC96501 RC965016 RC965032
Zinc sulfate 1.180 Specific Gravity for concentration of intestinal parasites, eggs, and cysts
Zinc sulphate


Formula: O₄SZn
MW: 161.45 g/mol
MDL Number: MFCD00011302
CAS Number: 7733-02-0
UN: 3077
ADR: 9,III
Order Now
Specification Test Results
Appearance Clear, colorless, odorless
Specific Gravity 1.175 ± 0.005 sp gr at 25.0 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty