Would you like to log in again?
Cadmium AAS Standards, RICCA Chemical Company
Supplier: RICCA Chemical Company
RACD1KH0-500N RACD1KN0-500N S2004000-500N RACD1KN0-100N RACD1KH0-100N
RCACD1KN-500EA 131.82 USD InStock
RCS2004000-5N RCACD1KN-500 RCACD1KH-100 RCACD1KN-100 RCACD1KH-500
Cadmium AAS Standards, RICCA Chemical Company
Standards Cationic Standards Cadmium Standards
1000 ppm is equivalernt to 1000 µg/ml
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty