Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-Camphene, natural isolate

Supplier: Ricca Chemical
Warning
Synonyms: (±)-3,3-Dimethyl-2-methylennorcamphaneCamphene(±)-2,2-Dimethyl-3-methylenebicyclo[2.2.1]heptane

RCCT9028-20C6 RCCT9028-750C6 RCCT9028-100C6
RCCT9028-20C6EA 44.15 USD InStock
RCCT9028-20C6 RCCT9028-750C6 RCCT9028-100C6
(±)-Camphene, natural isolate
(±)-Camphene


Formula: C₁₀H₁₆
MW: 136.24 g/mol
MDL Number: MFCD00066603
CAS Number: 79-92-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty