Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

β-Pinene, natural isolate

Supplier: Ricca Chemical
Danger
Synonyms: Nopinenebeta-PinenePseudopinene2(10)-PineneTerebenthene

RCCT9029-100C6 RCCT9029-750C6 RCCT9029-20C6
RCCT9029-20C6EA 21.19 USD InStock
RCCT9029-20C6 RCCT9029-100C6 RCCT9029-750C6
β-Pinene, natural isolate
β-Pinene


Formula: C₁₀H₁₆
MW: 136.23 g/mol
MDL Number: MFCD00064767
CAS Number: 127-91-3
UN: 2319
ADR: 3,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty