Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

β-Caryophyllene, natural isolate

Supplier: Ricca Chemical
Danger
Synonyms: (−)-β-Caryophyllene(1R,9S,E)-4,11,11-Trimethyl-8-methylenebicyclo[7.2.0]undec-4-ene(-)-trans-caryophyllene(1R,4E,9S)-4,11,11-trimethyl-8-methylene-bicyclo[7.2.0]undec-4-eneNSC 11906

RCCT9030-100C6 RCCT9030-20C6 RCCT9030-750C6
RCCT9030-750C6EA 165.98 USD InStock
RCCT9030-750C6 RCCT9030-100C6 RCCT9030-20C6
β-Caryophyllene, natural isolate
β-Caryophyllene


Formula: C₁₅H₂₄
MW: 204.36 g/mol
MDL Number: MFCD00075925
CAS Number: 87-44-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty