Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

α-Terpinene, natural isolate

Supplier: Ricca Chemical
Danger
Synonyms: 1-Isopropyl-4-methylcyclohexa-1,3-dienep-Mentha-1,3-diene

RCCT9032-750C6 RCCT9032-100C6 RCCT9032-20C6
RCCT9032-20C6EA 38.85 USD InStock
RCCT9032-20C6 RCCT9032-750C6 RCCT9032-100C6
α-Terpinene, natural isolate
α-Terpinene


Formula: C₁₀H₁₆
MW: 136.24 g/mol
MDL Number: MFCD00001534
CAS Number: 99-86-5
UN: 2319
ADR: 3,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty