Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(+/-)-ɑ-Bisabolol, natural isolate

Supplier: Ricca Chemical
Warning
Synonyms: rel-(R)-6-Methyl-2-((R)-4-methylcyclohex-3-en-1-yl)hept-5-en-2-ol6-Methyl-2-(4-methyl-3-cyclohexen-1-yl)-5-hepten-2-ol

RCCT9034-20C6 RCCT9034-100C6 RCCT9034-750C6
RCCT9034-100C6EA 353.15 USD InStock
RCCT9034-100C6 RCCT9034-750C6 RCCT9034-20C6
(+/-)-ɑ-Bisabolol, natural isolate
(+/-)-alpha-Bisabolol


Formula: C₁₅H₂₆O
MW: 222.37 g/mol
MDL Number: MFCD00129082
CAS Number: 515-69-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty