Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Myrcene, natural isolate

Supplier: Ricca Chemical
Danger

RCCT9085-20C6 RCCT9085-750C6 RCCT9085-100C6
RCCT9085-100C6EA 123.6 USD InStock
RCCT9085-100C6 RCCT9085-20C6 RCCT9085-750C6
Myrcene, natural isolate
Myrcene


Formula: C₁₀H₁₆
MDL Number: MFCD00008908
CAS Number: 123-35-3
UN: 2319
ADR: 3,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty