Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ocimene, natural isolate

Supplier: Ricca Chemical
Warning
Synonyms: 3,7-Dimethyl-1,3,6-octatrien3,7-Dimethyl-1,3,6-octatriene

RCCT9196-20C6 RCCT9196-100C6 RCCT9196-750C6
RCCT9196-750C6EA 2472 USD InStock
RCCT9196-750C6 RCCT9196-100C6 RCCT9196-20C6
Ocimene, natural isolate
Ocimene


Formula: C₁₀H₁₆
MW: 136.23 g/mol
MDL Number: MFCD00026437
CAS Number: 13877-91-3
UN: 2319
ADR: 3,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty