Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Citral, natural isolate

Supplier: Ricca Chemical
Warning
Synonyms: 3,7-Dimethylocta-2,6-dienal

RCCT9212-100C6 RCCT9212-750C6 RCCT9212-20C6
RCCT9212-20C6EA 28.26 USD InStock
RCCT9212-20C6 RCCT9212-750C6 RCCT9212-100C6
Citral, natural isolate
Citral


Formula: C₁₀H₁₆O
MW: 152.24 g/mol
MDL Number: MFCD00006997
CAS Number: 5392-40-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty